Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-chloro-4,5-difluorobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

31860-1G 31860-25G 31860-500MG 31860-5G
76940-036EA 334.19 USD InStock
76940-036 76940-038 76940-040 76940-042
Ethyl 2-chloro-4,5-difluorobenzoate ≥97%
Ethyl 2-chloro-4,5-difluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₇ClF₂O₂
MW: 220.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025300
CAS Number: 128800-37-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty