Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2,6-Difluoro-5-formylbenzoate ≥97%

Supplier: AOB CHEM USA
Warning

32219-1G 32219-25G 32219-500MG 32219-5G
76940-136EA 173.99 USD InStock
76940-136 76940-138 76940-140 76940-142
Methyl 2,6-Difluoro-5-formylbenzoate ≥97%
Methyl 2,6-Difluoro-5-formylbenzoate
Precaución: for Research Use Only


Formula: C₉H₆F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD30344687
CAS Number: 1993479-30-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty