Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoro-2-methoxy-3-methylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

32490-1G 32490-25G 32490-500MG 32490-5G
76940-242EA 350.1 USD InStock
76940-242 76940-244 76940-246 76940-248
6-Fluoro-2-methoxy-3-methylbenzoic acid ≥97%
6-Fluoro-2-methoxy-3-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22415158
CAS Number: 1427433-12-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty