Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-bromo-2,3-difluorobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

32517-1G 32517-25G 32517-5G
76940-254EA 94.43 USD InStock
76940-254 76940-256 76940-258
Methyl 4-bromo-2,3-difluorobenzoate ≥97%
Methyl 4-bromo-2,3-difluorobenzoate
Precaución: for Research Use Only


Formula: C₈H₅BrF₂O₂
MW: 251.025 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27578309
CAS Number: 1807244-07-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty