Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-bromo-2-chloropyridine-4-carboxylate ≥95%

Supplier: AOB CHEM USA
Warning

33490-1G 33490-25G 33490-500MG 33490-5G
76940-462EA 239.77 USD InStock
76940-462 76940-464 76940-466 76940-468
Methyl 3-bromo-2-chloropyridine-4-carboxylate ≥95%
Methyl 3-bromo-2-chloropyridine-4-carboxylate
Precaución: for Research Use Only


Formula: C₇H₅BrClNO₂
MW: 250.477 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698103
CAS Number: 1214385-62-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty