Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-fluoro-3-(trifluoromethoxy)phenol ≥97% research grade

Supplier: AOB CHEM USA
Warning

33661-1G 33661-25G 33661-500MG 33661-5G
76940-544EA 260.99 USD InStock
76940-544 76940-546 76940-548 76940-550
6-Bromo-2-fluoro-3-(trifluoromethoxy)phenol ≥97% research grade
6-Bromo-2-fluoro-3-(trifluoromethoxy)phenol


Formula: C₇H₃BrF₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD28740790
CAS Number: 1805580-01-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty