Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-bromo-2,6-difluorobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

33688-1G 33688-25G 33688-500MG 33688-5G
76940-556EA 153.84 USD InStock
76940-556 76940-558 76940-560 76940-562
Ethyl 3-bromo-2,6-difluorobenzoate ≥97%
Ethyl 3-bromo-2,6-difluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₇BrF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18917891
CAS Number: 1309933-04-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty