Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-(Trifluoromethyl)-1H-indazole ≥97% research grade

Supplier: AOB CHEM USA
Warning

34122-1G 34122-25G 34122-500MG 34122-5G
76940-642EA 213.25 USD InStock
76940-642 76940-644 76940-646 76940-648
7-(Trifluoromethyl)-1H-indazole ≥97% research grade
7-(Trifluoromethyl)-1H-indazole


Formula: C₈H₅F₃N₂
Storage Temperature: Refrigerator
MDL Number: MFCD09836171
CAS Number: 885694-00-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty