Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-fluoro-6-(trifluoromethyl)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

34987-1G 34987-25G 34987-500MG 34987-5G
76941-068EA 198.39 USD InStock
76941-068 76941-070 76941-072 76941-074
Methyl 2-fluoro-6-(trifluoromethyl)benzoate ≥97%
Methyl 2-fluoro-6-(trifluoromethyl)benzoate
Precaución: for Research Use Only


Formula: C₉H₆F₄O₂
MW: 222.136 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06203704
CAS Number: 153556-50-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty