Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2,4-dichloro-3-hydroxybenzoate ≥97%

Supplier: AOB CHEM USA
Warning

35060-1G 35060-500MG 35060-5G
76941-112EA 587.74 USD InStock
76941-112 76941-114 76941-116
Methyl 2,4-dichloro-3-hydroxybenzoate ≥97%
Methyl 2,4-dichloro-3-hydroxybenzoate
Precaución: for Research Use Only


Formula: C₈H₆Cl₂O₃
MW: 221.037 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28786464
CAS Number: 129788-30-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty