Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-bromo-6-methoxyisonicotinate ≥97%

Supplier: AOB CHEM USA
Warning

35571-1G 35571-25G 35571-500MG 35571-5G
76941-264EA 381.93 USD InStock
76941-264 76941-266 76941-268 76941-270
Methyl 2-bromo-6-methoxyisonicotinate ≥97%
Methyl 2-bromo-6-methoxyisonicotinate
Precaución: for Research Use Only


Formula: C₈H₈BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD18258550
CAS Number: 853030-09-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty