Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isopropyl 4-bromo-3-methylbenzoate ≥97%

Supplier: AOB CHEM USA
Warning

36028-25G 36028-5G
76941-352EA 456.19 USD InStock
76941-352 76941-354
Isopropyl 4-bromo-3-methylbenzoate ≥97%
Isopropyl 4-bromo-3-methylbenzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₃BrO₂
MW: 257.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26802269
CAS Number: 360773-73-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty