Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-fluoro-3-formylbenzoate ≥97%

Supplier: AOB CHEM USA
Warning

36123-1G 36123-25G 36123-500MG 36123-5G
76941-370EA 147.47 USD InStock
76941-370 76941-372 76941-374 76941-376
Methyl 2-fluoro-3-formylbenzoate ≥97%
Methyl 2-fluoro-3-formylbenzoate
Precaución: for Research Use Only


Formula: C₉H₇FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD24447977
CAS Number: 1262419-96-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty