Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-(benzyloxy)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

36128-1G 36128-25G 36128-500MG 36128-5G
76941-382EA 118.83 USD InStock
76941-382 76941-384 76941-386 76941-388
Ethyl 3-(benzyloxy)benzoate ≥97%
Ethyl 3-(benzyloxy)benzoate
Precaución: for Research Use Only


Formula: C₁₆H₁₆O₃
MW: 256.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204302
CAS Number: 63888-94-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty