Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-bromo-5-iodobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

36356-1G 36356-25G 36356-5G
76941-464EA 103.97 USD InStock
76941-464 76941-466 76941-468
Ethyl 3-bromo-5-iodobenzoate ≥97%
Ethyl 3-bromo-5-iodobenzoate
Precaución: for Research Use Only


Formula: C₉H₈BrIO₂
MW: 354.967 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658440
CAS Number: 186772-44-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty