Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-hydroxy-5-methylbenzoate ≥97%

Supplier: AOB CHEM USA
Warning

36578-1G 36578-25G 36578-500MG 36578-5G
76941-520EA 118.83 USD InStock
76941-520 76941-522 76941-524 76941-526
Ethyl 3-hydroxy-5-methylbenzoate ≥97%
Ethyl 3-hydroxy-5-methylbenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD18396519
CAS Number: 1126430-75-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty