Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-chloro-2-fluorobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

36774-1G 36774-25G 36774-5G
76941-580EA 62.6 USD InStock
76941-580 76941-582 76941-584
Ethyl 5-chloro-2-fluorobenzoate ≥97%
Ethyl 5-chloro-2-fluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD06205125
CAS Number: 773139-56-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty