Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-(benzyloxy)-3-chlorobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

37207-1G 37207-25G 37207-500MG 37207-5G
76941-752EA 354.35 USD InStock
76941-752 76941-754 76941-756 76941-758
tert-Butyl 4-(benzyloxy)-3-chlorobenzoate ≥97%
tert-Butyl 4-(benzyloxy)-3-chlorobenzoate
Precaución: for Research Use Only


Formula: C₁₈H₁₉ClO₃
MW: 318.795 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32632163
CAS Number: 2379322-84-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty