Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isopropyl 2-bromo-5-(trifluoromethyl)benzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

37584-25G 37584-5G
76941-898EA 346.92 USD InStock
76941-898 76941-900
Isopropyl 2-bromo-5-(trifluoromethyl)benzoate ≥97% research grade
Isopropyl 2-bromo-5-(trifluoromethyl)benzoate


Formula: C₁₁H₁₀BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD23581985
CAS Number: 1506090-22-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty