Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 5-bromo-6-methylpicolinate ≥97% research grade

Supplier: AOB CHEM USA
Warning

37639-1G 37639-25G 37639-500MG 37639-5G
76941-956EA 260.99 USD InStock
76941-956 76941-958 76941-960 76941-962
tert-Butyl 5-bromo-6-methylpicolinate ≥97% research grade
tert-Butyl 5-bromo-6-methylpicolinate


Formula: C₁₁H₁₄BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD32632251
CAS Number: 2404734-24-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty