Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-(benzyloxy)-3,5-difluorobenzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

37986-1G 37986-25G 37986-500MG 37986-5G
76942-036EA 376.62 USD InStock
76942-036 76942-038 76942-040 76942-042
Methyl 4-(benzyloxy)-3,5-difluorobenzoate ≥97% research grade
Methyl 4-(benzyloxy)-3,5-difluorobenzoate


Formula: C₁₅H₁₂F₂O₃
MW: 278.251 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28142132
CAS Number: 1431533-45-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty