Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2,4-bis(trifluoromethyl)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

38657-1G 38657-25G 38657-500MG 38657-5G
76942-314EA 153.84 USD InStock
76942-314 76942-316 76942-318 76942-320
Ethyl 2,4-bis(trifluoromethyl)benzoate ≥97%
Ethyl 2,4-bis(trifluoromethyl)benzoate


Formula: C₁₁H₈F₆O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06204429
CAS Number: 773135-00-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty