Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Bromo-6-methoxy-1H-indazole ≥97% research grade

Supplier: AOB CHEM USA
Warning

38686-1G 38686-25G 38686-500MG 38686-5G
76942-330EA 406.33 USD InStock
76942-330 76942-332 76942-334 76942-336
7-Bromo-6-methoxy-1H-indazole ≥97% research grade
7-Bromo-6-methoxy-1H-indazole


Formula: C₈H₇BrN₂O
MW: 227.058 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22380507
CAS Number: 1374651-66-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty