Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-bromo-4-methylnicotinate ≥97% research grade

Supplier: AOB CHEM USA
Warning

45857-1G 45857-25G 45857-500MG 45857-5G
76942-476EA 287.51 USD InStock
76942-476 76942-478 76942-480 76942-482
Ethyl 5-bromo-4-methylnicotinate ≥97% research grade
Ethyl 5-bromo-4-methylnicotinate


Formula: C₉H₁₀BrNO₂
MW: 244.085 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28145641
CAS Number: 1428651-84-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty