Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl biphenyl-2-carboxylate ≥97% research grade

Supplier: AOB CHEM USA
Warning

45982-1G 45982-25G 45982-500MG 45982-5G
76942-530EA 221.73 USD InStock
76942-530 76942-532 76942-534 76942-536
Ethyl biphenyl-2-carboxylate ≥97% research grade
Ethyl biphenyl-2-carboxylate


Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00671951
CAS Number: 19926-49-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty