Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-bromo-6-chloronicotinate ≥97% research grade

Supplier: AOB CHEM USA
Warning

48237-1G 48237-25G 48237-500MG 48237-5G
76942-742EA 144.29 USD InStock
76942-742 76942-744 76942-746 76942-748
Ethyl 5-bromo-6-chloronicotinate ≥97% research grade
Ethyl 5-bromo-6-chloronicotinate


Formula: C₈H₇BrClNO₂
MW: 264.504 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698309
CAS Number: 952063-30-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty