Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-hydroxy-5-(trifluoromethyl)benzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

49041-1G 49041-25G 49041-5G
76942-822EA 62.6 USD InStock
76942-822 76942-824 76942-826
Methyl 2-hydroxy-5-(trifluoromethyl)benzoate ≥97% research grade
Methyl 2-hydroxy-5-(trifluoromethyl)benzoate


Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399428
CAS Number: 1261478-26-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty