Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-chloro-3-ethoxy-4-fluorobenzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

52467-1G 52467-25G 52467-500MG 52467-5G
76943-054EA 240.83 USD InStock
76943-054 76943-056 76943-058 76943-060
Methyl 2-chloro-3-ethoxy-4-fluorobenzoate ≥97% research grade
Methyl 2-chloro-3-ethoxy-4-fluorobenzoate


Formula: C₁₀H₁₀ClFO₃
Storage Temperature: Ambient
CAS Number: 2384943-78-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty