Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-(benzylthio)benzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

53063-1G 53063-25G 53063-500MG 53063-5G
76943-082EA 144.29 USD InStock
76943-082 76943-084 76943-086 76943-088
Methyl 4-(benzylthio)benzoate ≥97% research grade
Methyl 4-(benzylthio)benzoate


Formula: C₁₅H₁₄O₂S
MW: 258.335 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28359625
CAS Number: 26960-78-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty