Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Cyanophenyl)acetic acid ethyl ester ≥95%

Supplier: AOB CHEM USA
Warning

57760-25G 57760-5G
76943-418EA 183.54 USD InStock
76943-418 76943-420
(4-Cyanophenyl)acetic acid ethyl ester ≥95%
(4-Cyanophenyl)acetic acid ethyl ester
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₂
MW: 189.21
Storage Temperature: Refrigerator
MDL Number: MFCD09953133
CAS Number: 1528-41-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty