Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoro-3-methyl-1H-indazole ≥97%

Supplier: AOB CHEM USA
Warning

61772-1G 61772-25G 61772-500MG 61772-5G
76943-650EA 260.99 USD InStock
76943-650 76943-652 76943-654 76943-656
6-Fluoro-3-methyl-1H-indazole ≥97%
6-Fluoro-3-methyl-1H-indazole


Formula: C₈H₇FN₂
Storage Temperature: Ambient
MDL Number: MFCD08692363
CAS Number: 159305-16-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty