Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-((4-fluorobenzyl)oxy)benzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

62509-1G 62509-500MG 62509-5G
76943-686EA 1434.34 USD InStock
76943-686 76943-688 76943-690
Methyl 3-((4-fluorobenzyl)oxy)benzoate ≥97% research grade
Methyl 3-((4-fluorobenzyl)oxy)benzoate


Formula: C₁₅H₁₃FO₃
MW: 260.2603 g/mol
Storage Temperature: Ambient
CAS Number: 381234-84-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty