Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,5-Trifluorothioanisole ≥97%

Supplier: AOB CHEM USA
Warning

63261-1G 63261-25G 63261-500MG 63261-5G
76943-712EA 195.21 USD InStock
76943-712 76943-714 76943-716 76943-718
2,4,5-Trifluorothioanisole ≥97%
2,4,5-Trifluorothioanisole
Precaución: for Research Use Only


Formula: C₇H₅F₃S
MW: 178.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12026061
CAS Number: 54378-74-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty