Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-cyano-5-methylbenzoate ≥95% research grade

Supplier: AOB CHEM USA
Warning

75354-1G 75354-25G 75354-500MG 75354-5G
76943-846EA 183.54 USD InStock
76943-846 76943-848 76943-850 76943-852
Methyl 3-cyano-5-methylbenzoate ≥95% research grade
Methyl 3-cyano-5-methylbenzoate


Formula: C₁₀H₉NO₂
MW: 175.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11007826
CAS Number: 1082041-26-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty