Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxy-2H-chromen-2-one ≥97%

Supplier: AOB CHEM USA
Warning

84184-1G 84184-25G 84184-500MG 84184-5G
76944-078EA 236.59 USD InStock
76944-078 76944-080 76944-082 76944-084
6-Methoxy-2H-chromen-2-one ≥97%
6-Methoxy-2H-chromen-2-one


Formula: C₁₀H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD00016707
CAS Number: 17372-53-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty