Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥95%

Supplier: AOB CHEM USA
Warning

99091-1G 99091-25G 99091-5G
76944-168EA 79.57 USD InStock
76944-168 76944-170 76944-172
Methyl-4-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥95%
Methyl 4-(trifluoromethyl)benzo[b]thiophene-2-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₇F₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD09027107
CAS Number: 146137-87-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty