Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichlorobenzothiazole 97%

Supplier: AMBEED, INC
Warning

A209274-1G A209274-250MG A209274-25G A209274-5G A209274-10G
76975-476EA 6 USD InStock
76975-476 76975-478 76975-480 76975-482 77671-858
2,6-Dichlorobenzothiazole 97%
2,6-Dichlorobenzothiazole
Precaución: for Research Use Only


Formula: C₇H₃Cl₂NS
MDL Number: MFCD00044101
CAS Number: 3622-23-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty