Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Ethoxy-1H-indole 98%

Supplier: AMBEED, INC
Warning

A194181-100MG A194181-1G A194181-250MG A194181-25G A194181-5G
76974-978EA 29 USD InStock
76974-978 76974-980 76974-982 76974-984 76974-986
6-Ethoxy-1H-indole 98%
6-Ethoxy-1H-indole
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO
MDL Number: MFCD00051498
CAS Number: 37865-86-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty