Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-Hydroxyethyl)phthalimide 98%

Supplier: AMBEED, INC
Warning

A144263-1000G A144263-100G A144263-10G A144263-25G A144263-500G
76972-740EA 49 USD InStock
76972-740 76972-742 76972-744 76972-746 76972-748
N-(2-Hydroxyethyl)phthalimide 98%
N-(2-Hydroxyethyl)phthalimide
Precaución: for Research Use Only


Formula: C₁₀H₉NO₃
MDL Number: MFCD00005903
CAS Number: 3891-07-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty