Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Chlorophenyl)pyrrolidine 98%

Supplier: AMBEED, INC
Warning

A817300-100MG A817300-1G A817300-250MG A817300-5G
76981-712EA 26 USD InStock
76981-712 76981-714 76981-716 76981-718
2-(4-Chlorophenyl)pyrrolidine 98%
2-(4-Chlorophenyl)pyrrolidine
Precaución: for Research Use Only


Formula: C₁₀H₁₂ClN
MDL Number: MFCD01631838
CAS Number: 38944-14-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty