Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-fluorobenzoyl chloride 98%

Supplier: AMBEED, INC
Danger

A293033-5G A293033-25G
76977-616EA 18 USD InStock
76977-616 77672-492
5-Chloro-2-fluorobenzoyl chloride 98%
5-Chloro-2-fluorobenzoyl chloride
Precaución: for Research Use Only


Formula: C₇H₃Cl₂FO
MW: 193
Density: 1.459
MDL Number: MFCD01631433
CAS Number: 394-29-6
UN: 3265
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty