Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-methoxybenzoic acid 95%

Supplier: AMBEED, INC
Warning

A182357-100G A182357-1G A182357-25G A182357-5G
76974-482EA 317 USD InStock
76974-482 76974-484 76974-486 76974-488
4-Fluoro-2-methoxybenzoic acid 95%
4-Fluoro-2-methoxybenzoic acid
Precaución: for Research Use Only


Formula: C₈H₇FO₃
MW: 170.14
MDL Number: MFCD00673010
CAS Number: 395-82-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty