Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Acetyl-2-pyridine carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A762301-100MG A762301-1G A762301-250MG
76981-430EA 71 USD InStock
76981-430 76981-432 76981-434
3-Acetyl-2-pyridine carboxylic acid 95%
3-Acetyl-2-pyridine carboxylic acid
Precaución: for Research Use Only


Formula: C₈H₇NO₃
MDL Number: MFCD16140257
CAS Number: 716362-04-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty