Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-methoxypyrazine 98%

Supplier: AMBEED, INC
Warning

A277478-100G A277478-10G A277478-1G A277478-25G A277478-5G
76977-312EA 255 USD InStock
76977-312 76977-314 76977-316 76977-318 76977-320
2-Chloro-3-methoxypyrazine 98%
2-Chloro-3-methoxypyrazine
Precaución: for Research Use Only


Formula: C₅H₅ClN₂O
MDL Number: MFCD02684324
CAS Number: 40155-28-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty