Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Fluoroacetophenone 98%

Supplier: AMBEED, INC
Warning

A172053-100G A172053-10G A172053-25G A172053-500G
76974-054EA 11 USD InStock
76974-054 76974-056 76974-058 76974-060
4'-Fluoroacetophenone 98%
4'-Fluoroacetophenone
Precaución: for Research Use Only


Formula: C₈H₇FO
MW: 138.14 g/mol
Boiling Pt: 196
MDL Number: MFCD00000354
CAS Number: 403-42-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty