Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-hydroxybenzonitrile 98%

Supplier: AMBEED, INC
Warning

A411028-100G A411028-10G A411028-1G A411028-25G A411028-5G
76979-132EA 82 USD InStock
76979-132 76979-134 76979-136 76979-138 76979-140
3-Fluoro-4-hydroxybenzonitrile 98%
3-Fluoro-4-hydroxybenzonitrile
Precaución: for Research Use Only


Formula: C₇H₄FNO
MDL Number: MFCD00215834
CAS Number: 405-04-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty