Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-N-benzylpropanamide hydrochloride 95%

Supplier: AMBEED, INC
Warning

A513749-1G A513749-5G
76979-934EA 82 USD InStock
76979-934 76979-936
2-Amino-N-benzylpropanamide hydrochloride 95%
2-Amino-N-benzylpropanamide hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₅ClN₂O
MW: 214.7 g/mol
MDL Number: MFCD13195941
CAS Number: 1214706-98-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty