Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-Acetamido-2-methoxybenzoate 95%

Supplier: AMBEED, INC
Warning

A148502-100G A148502-10G A148502-1G A148502-250MG A148502-25G A148502-500G A148502-5G
76972-966EA 35 USD InStock
76972-966 76972-968 76972-970 76972-972 76972-974 76972-976 76972-978
Methyl 4-Acetamido-2-methoxybenzoate 95%
Methyl 4-Acetamido-2-methoxybenzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₃NO₄
MDL Number: MFCD00065258
CAS Number: 4093-29-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty