Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methyl-4-nitrophthalimide 97%

Supplier: AMBEED, INC
Warning

A117896-100G A117896-10G A117896-25G A117896-500G
76971-334EA 20 USD InStock
76971-334 76971-336 76971-338 76971-340
N-Methyl-4-nitrophthalimide 97%
N-Methyl-4-nitrophthalimide
Precaución: for Research Use Only


Formula: C₉H₆N₂O₄
MW: 206.16 g/mol
MDL Number: MFCD00454263
CAS Number: 41663-84-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty