Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ɑ-[2-(Methylamino)ethyl]benzyl alcohol 97%

Supplier: AMBEED, INC
Warning

A208236-1000G A208236-100G A208236-25G A208236-500G
76975-418EA 270 USD InStock
76975-418 76975-420 76975-422 76975-424
ɑ-[2-(Methylamino)ethyl]benzyl alcohol 97%
alpha-[2-(Methylamino)ethyl]benzyl alcohol
Precaución: for Research Use Only


Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
MDL Number: MFCD00674078
CAS Number: 42142-52-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty